missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Membrin Antibody (8G6), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42976
This item is not returnable.
View return policy
Description
Membrin Monoclonal antibody specifically detects Membrin in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| Membrin | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Immunohistochemistry, Western Blot | |
| 8G6 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunohistochemistry 1:20 - 1:200 | |
| golgi SNAP receptor complex member 2, GS27Bos1,27 kDa Golgi SNARE protein, membrin | |
| golgi SNAP receptor complex member 2 - (Uniprot# O14653) | |
| 100 μg | |
| Membrane Trafficking and Chaperones | |
| 9570 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction