missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Heparanase/HPSE Antibody (0C0), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42982
This item is not returnable.
View return policy
Description
Heparanase/HPSE Monoclonal antibody specifically detects Heparanase/HPSE in Human samples. It is validated for ELISA,Western Blot,Immunocytochemistry/ Immunofluorescence
Specifications
| Heparanase/HPSE | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunocytochemistry/Immunofluorescence | |
| 0C0 | |
| Western Blot 1:500 - 1:2000, ELISA, Immunocytochemistry/ Immunofluorescence 1:50 - 1:500 | |
| EC 3.2, endo-glucoronidase, HEP, heparanase, Heparanase-1, Hpa1, HPAheparanase exon 9 and 10 deletion, HPR1, HPSE1heparanase-1, HSE1heparanase exon10-deletion | |
| heparanase - (Uniprot# Q9Y251) | |
| 100 μg | |
| Pancreatic Stem Cell Markers, Stem Cell Markers | |
| 10855 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction