missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTO1 Antibody (9H4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-42979
This item is not returnable.
View return policy
Description
GSTO1 Monoclonal antibody specifically detects GSTO1 in Human,Mouse,Rat samples. It is validated for ELISA,Immunohistochemistry,Western Blot
Specifications
| GSTO1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.3, 50% glycerol | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 9446 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG2a |
| ELISA, Immunohistochemistry, Western Blot | |
| 9H4 | |
| Western Blot 1:1000 - 1:4000, ELISA, Immunohistochemistry 1:50 - 1:200 | |
| DKFZp686H13163, EC 2.5.1.18, glutathione S-transferase omega 1, glutathione S-transferase omega 1-1, glutathione S-transferase omega-1, glutathione-S-transferase like, GSTO-1, GSTTLp28, P28 | |
| glutathione S-transferase omega 1 - (Uniprot# P78417) | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction