missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DNA-RNA Hybrid Antibody (S9.6), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45094
This item is not returnable.
View return policy
Description
DNA-RNA Hybrid Monoclonal antibody specifically detects DNA-RNA Hybrid in All samples. It is validated for ChIP Assay,Flow Cytometry,Immunoprecipitation,Immunocytochemistry/ Immunofluorescence
Specifications
| DNA-RNA Hybrid | |
| Monoclonal | |
| 1 mg/mL | |
| 10 mM PBS | |
| DNA-RNA heteropolymer duplex prepared by transcription of phi X174 single-stranded DNA with DNA-dependent RNA polymerase | |
| 100 μg | |
| Primary | |
| Store at -20 to -84°C. Avoid freeze-thaw cycles. | |
| Purified |
| ChIP Assay, Flow Cytometry, Immunoprecipitation, Immunocytochemistry/Immunofluorescence | |
| S9.6 | |
| Unconjugated | |
| Mouse | |
| Protein A purified | |
| RUO | |
| All species | |
| Antibody | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction