missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human RBPMS Antibody, Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB11670-100
This item is not returnable.
View return policy
Description
RBPMS Monoclonal antibody specifically detects RBPMS in Human samples. It is validated for Western Blot,Immunohistochemistry
Specifications
| RBPMS | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2μm filtered solution in PBS with Trehalose. | |
| FLJ32971, Heart and RRM expressed sequence, Hermes, HERMESRNA-binding protein with multiple splicing, RBPMS, RBP-MS, RNA binding protein with multiple splicing | |
| Synthetic Peptide, Accession # Q93062 | |
| 100 μg | |
| Primary | |
| Reconstitute lyophilized material at 0.2 mg/mL in sterile PBS. For liquid material, refer to CoA for concentration. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 1090828 | |
| Western Blot 2 μg/mL, Immunohistochemistry 3-25 μg/mL | |
| Q93062 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 11030 | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction