missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Semaphorin 3E Alexa Fluor 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC32391S-100UG
This item is not returnable.
View return policy
Description
Semaphorin 3E Monoclonal specifically detects Semaphorin 3E in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Semaphorin 3E | |
| Monoclonal | |
| 0.2 mg/mL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| O15041 | |
| SEMA3E | |
| Mouse myeloma cell line NS0-derived recombinant human Semaphorin 3E Thr25-Ser775 (Arg557Ala and Arg560Ala) Accession # O15041 | |
| RUO | |
| 9723 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| 400513 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| (semaphorin) 3E, coll-5, KIAA0331M-SEMAH, sema domain, immunoglobulin domain (Ig), short basic domain, secreted, Sema3E, SEMAH, SEMAHM-SemaK, semaphorin-3E | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Semaphorin 3E in direct ELISAs. In direct ELISAs, approximately 15% cross-reactivity with recombinant human (rh) Semaphorin 3B is observed and no cross-reactivity with rhSemaphorin 6A is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction