missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human FCRN Alexa Fluor 488-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC8639G-100UG
This item is not returnable.
View return policy
Description
FCRN/FCGRT Monoclonal specifically detects FCRN/FCGRT in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| FCRN/FCGRT | |
| Monoclonal | |
| Alexa Fluor 488 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| alpha-chain, Fc fragment of IgG, receptor, transporter, alpha, FCGRT, FcRn, FcRn alpha chain, FCRNimmunoglobulin receptor, intestinal, heavy chain, IgG Fc fragment receptor transporter alpha chain, IgG receptor FcRn large subunit p51, major histocompatibility complex class I-like Fc receptor, Neonatal Fc receptor, neonatal Fc-receptor for Ig | |
| Chinese hamster ovary cell line CHO-derived recombinant human FCRN Ala24-Ser297 Accession # P55899 | |
| RUO | |
| 2217 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 937508 | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| P55899 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human FCRN in direct ELISAs. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction