missing translation for 'onlineSavingsMsg'
Learn More

Vioxx, TRC

Product Code. 30008259
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008259 100 mg 100mg
30011956 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30008259 Supplier Toronto Research Chemicals Supplier No. TRCV670000100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 162011-90-7
Molecular Formula C17 H14 O4 S
Molecular Weight (g/mol) 314.36
IUPAC Name 3-(4-methylsulfonylphenyl)-4-phenyl-2H-furan-5-one
SMILES CS(=O)(=O)c1ccc(cc1)C2=C(C(=O)OC2)c3ccccc3

Specifications

Quantity 100 mg
Synonym 4-[4-(Methylsulfonyl)phenyl]-3-phenyl-2(5H)-furanone, 3-(4-Methanesulfonylphenyl)-2-phenyl-2-buten-4-olide, 3-Phenyl-4-[4-(Methylsulfonyl)phenyl]-2(5H)-furanone, 4-[4-(Methanesulfonyl)phenyl]-3-phenyl-5H-furan-2-one, Ceeoxx, Ceoxx, MK 0966, MK 966, Rhuma-cure, Rofecoxib, Romacox, Vioxx
InChI Formula InChI=1S/C17H14O4S/c1-22(19,20)14-9-7-12(8-10-14)15-11-21-17(18)16(15)13-5-3-2-4-6-13/h2-10H,11H2,1H3
Formula Weight 314.0613
Percent Purity >95%
Chemical Name or Material Vioxx
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.