accessibility menu, dialog, popup

UserName

CAS RN 162011-90-7

CH3SOOOO

CAS RN 162011-90-7

IUPAC Name: 4-(4-methanesulfonylphenyl)-3-phenyl-2,5-dihydrofuran-2-one
Synonyms: rofecoxib vioxx ceoxx
Molecular Weight (g/mol): 314.36
Molecular Formula: C17H14O4S
InChi Key: RZJQGNCSTQAWON-UHFFFAOYSA-N
SMILES: CS(=O)(=O)C1=CC=C(C=C1)C1=C(C(=O)OC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results