missing translation for 'onlineSavingsMsg'
Learn More

Trazodone.HCl, Lipomed

Product Code. 30460818
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30460818

Brand: Lipomed LPMTZD1373HC100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.

Trazodone.HCl
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Trazodone Hydrochloride
CAS 25332-39-2
Analyte or Component Names Trazodone Hydrochloride
Shipping Condition Room Temperature
Molecular Formula C19 H22 Cl N5 O . Cl H
Synonym 1,2,4-Triazolo[4,3-a]pyridin-3(2 H)-one, 2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-, hydrochloride (1:1), 1,2,4-Triazolo[4,3-a]pyridin-3(2 H)-one, 2-[3-[4-(3-chlorophenyl)-1-piperazinyl]propyl]-, monohydrochloride (9 CI), s-Triazolo[4,3-a]pyridin-3(2 H)-one, 2-[3-[4-(m-chlorophenyl)-1-piperazinyl]propyl]-, monohydrochloride (8 CI), AF 1161, Bimaran, Desyrel, KB 831, Mesyrel, Molipaxin, Pragmazone, Thombran, Tombran, Trazodone hydrochloride, Trazolan, Trittico
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Cl.Clc1cccc(c1)N2CCN(CCCN3N=C4C=CC=CN4C3=O)CC2
IUPAC Name 2-[3-[4-(3-chlorophenyl)piperazin-1-yl]propyl]-[1,2,4 ]triazolo[4,3-a]pyridin-3-one;hydrochloride
Industry Type Pharmaceutical
Molecular Weight (g/mol) 408.32
Quantity 100 mg
Formula Weight 407.128 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.