missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pridinol Hydrochloride, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Pridinol Hydrochloride
Specifications
Specifications
| Chemical Name or Material | Pridinol Hydrochloride |
| CAS | 968-58-1 |
| Analyte or Component Names | Pridinol Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H25 N O . Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | Parks 12, Pridinol hydrochloride, Ridinol, alpha,alpha-Diphenyl-1-piperidinepropanol Hydrochloride (1:1), 1-Piperidinepropanol, alpha,alpha-diphenyl-, hydrochloride (6CI,7CI,8CI,9CI), alpha,alpha-Diphenyl-1-piperidinepropanol hydrochloride |
| InChI Formula | InChI=1S/C20H25NO.ClH/c22-20(18-10-4-1-5-11-18,19-12-6-2-7-13-19)14-17-21-15-8-3-9-16-21;/h1-2,4-7,10-13,22H,3,8-9,14-17H2;1H |
| SMILES | Cl.OC(CCN1CCCCC1)(c2ccccc2)c3ccccc3 |
| IUPAC Name | 1,1-diphenyl-3-piperidin-1-ylpropan-1-ol;hydrochloride |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction