missing translation for 'onlineSavingsMsg'
Learn More

Nicoboxil, Mikromol™

Product Code. 17932936
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17932936

Brand: Mikromol MM3800.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nicoboxil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nicoboxil
CAS 13912-80-6
Analyte or Component Names Nicoboxil
Shipping Condition Room Temperature
Molecular Formula C12H17NO3
Certifications/Compliance ISO 17034
Synonym 3-Pyridinecarboxylic acid 2-butoxyethyl ester, Nicotinic acid, 2-butoxyethyl ester (6CI,7CI,8CI), Ethanol, 2-butoxy-, nicotinate (8CI), 2-Butoxyethyl nicotinate, Nicoboxil, Nicotinic acid beta-butoxyethyl ester, beta-Butoxyethyl nicotinate
InChI Formula InChI=1S/C12H17NO3/c1-2-3-7-15-8-9-16-12(14)11-5-4-6-13-10-11/h4-6,10H,2-3,7-9H2,1H3
SMILES CCCCOCCOC(=O)c1cccnc1
IUPAC Name 2-butoxyethyl pyridine-3-carboxylate
Molecular Weight (g/mol) 223.27
Quantity 100 mg
Formula Weight 223.1208 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.