missing translation for 'onlineSavingsMsg'
Learn More

Nandrolone Decanoate, Mikromol™

Product Code. 17970396
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17970396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17970396 Supplier Mikromol Supplier No. MM0982.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Nandrolone Decanoate
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Nandrolone Decanoate
CAS 360-70-3
Analyte or Component Names Nandrolone Decanoate
Shipping Condition Ice pack (-20°C)
Molecular Formula C28 H44 O3
Certifications/Compliance ISO 17025
Synonym Nandrolone Decanoate, Estr-4-en-3-one, 17-[(1-oxodecyl)oxy]-, (17beta)-, Estr-4-en-3-one, 17beta-hydroxy-, decanoate (6CI,7CI,8CI), Decanoic acid, ester with 17beta-hydroxyestr-4-en-3-one (8CI), 17beta-Hydroxy-19-norandrost-4-en-3-one 17-decanoate, 17beta-Hydroxyestr-4-en-3-one 17-decanoate, 17beta-Hydroxyestr-4-en-3-one decanoate, 17beta-Nandrolone laureate, 17beta-Nortestosterone laureate, 19-Nor-17beta-hydroxy-3-ketoandrost-4-ene 17-decanoate, 19-Norandrostenolone decanoate, 19-Nortestosterone 17beta-decanoate, 19-Nortestosterone decanoate, Anabolicum, Anabolin Forte, Deca-Durabol, Deca-Durabolin, Deca-Hybolin, Decanofort, Extraboline, Hybolin decanoate, Nandrobolic LA, Norandrostenolone decanoate, Nortestosterone decanoate, Retabolil
InChI Formula InChI=1S/C28H44O3/c1-3-4-5-6-7-8-9-10-27(30)31-26-16-15-25-24-13-11-20-19-21(29)12-14-22(20)23(24)17-18-28(25,26)2/h19,22-26H,3-18H2,1-2H3/t22-,23+,24+,25-,26-,28-/m0/s1
SMILES CCCCCCCCCC(=O)O[C@H]1CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C
IUPAC Name [(8R,9S,10R,13S,14S,17S)-13-methyl-3-oxo-2,6,7,8,9,10,11,12,14,15,16,17-dodecahydro-1H-cyclopenta[a]phenanthren-17-yl] decanoate
Molecular Weight (g/mol) 428.647
Quantity 100 mg
Formula Weight 428.329 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.