missing translation for 'onlineSavingsMsg'
Learn More

Glycopyrronium Bromide, Mikromol™

Product Code. 17911836
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 17911836

Brand: Mikromol MM0834.00

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

Glycopyrronium Bromide
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glycopyrronium Bromide
CAS 51186-83-5
Analyte or Component Names Glycopyrronium Bromide
Shipping Condition Room Temperature
Molecular Formula C19 H28 N O3 . Br
Certifications/Compliance ISO 17034
Synonym Glycopyrronium Bromide, Pyrrolidinium, 3-[[(2R)-2-cyclopentyl-2-hydroxy-2-phenylacetyl]oxy]-1,1-dimethyl-, bromide (1:1), (3S)-rel-, Pyrrolidinium, 3-[(cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethyl-, bromide, (R*,S*)-(±)-, Pyrrolidinium, 3-[[(2R)-cyclopentylhydroxyphenylacetyl]oxy]-1,1-dimethyl-, bromide, (3S)-rel- (9CI), (3S)-rel-3-[[(2R)-2-Cyclopentyl-2-hydroxy-2-phenylacetyl]oxy]-1,1-dimethylpyrrolidinium bromide (1:1), (R*,S*)-3-[(Cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethylpyrrolidinium bromide, (R*,S*)-(±)-3-[(Cyclopentylhydroxyphenylacetyl)oxy]-1,1-dimethylpyrrolidinium bromide, erythro-Glycopyrronium bromide
InChI Formula InChI=1S/C19H28NO3.BrH/c1-20(2)13-12-17(14-20)23-18(21)19(22,16-10-6-7-11-16)15-8-4-3-5-9-15;/h3-5,8-9,16-17,22H,6-7,10-14H2,1-2H3;1H/q+1;/p-1/t17-,19-;/m0./s1
SMILES [Br-].C[N+]1(C)CC[C@@H](C1)OC(=O)[C@@](O)(C2CCCC2)c3ccccc3
IUPAC Name [(3S)-1,1-dimethylpyrrolidin-1-ium-3-yl] (2R)-2-cyclopentyl-2-hydroxy-2-phenylacetate;bromide
Molecular Weight (g/mol) 398.33
Quantity 100 mg
Formula Weight 398.3345 g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.