missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Etodolac Ethyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
Etodolac Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Etodolac Ethyl Ester |
| CAS | 200880-23-5 |
| Analyte or Component Names | Etodolac Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C19 H25 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Etodolac Ethyl Ester, 1,8-Diethyl-1,3,4,9-tetrahydro-pyrano[3,4-b]indole-1-acetic acid ethyl ester |
| InChI Formula | InChI=1S/C19H25NO3/c1-4-13-8-7-9-14-15-10-11-23-19(5-2,12-16(21)22-6-3)18(15)20-17(13)14/h7-9,20H,4-6,10-12H2,1-3H3 |
| SMILES | CCOC(=O)CC1(CC)OCCc2c1[nH]c3c(CC)cccc23 |
| IUPAC Name | ethyl 2-(1,8-diethyl-4,9-dihydro-3H-pyrano[3,4-b]indol-1-yl)acetate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction