missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Diclofenac 1,2-Propylene Glycol Ester (Mixture of Isomers), Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Diclofenac 1,2-Propylene Glycol Ester (Mixture of Isomers)
Specifications
Specifications
| Chemical Name or Material | Diclofenac 1,2-Propylene Glycol Ester (Mixture of Isomers) |
| Analyte or Component Names | Diclofenac 1,2-Propylene Glycol Ester (Mixture of Isomers) |
| Shipping Condition | Room Temperature |
| Molecular Formula | 2 C17 H17 Cl2 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Diclofenac 1,2-Propylene Glycol Ester (Mixture of Isomers) |
| InChI Formula | InChI=1S/2C17H17Cl2NO3/c1-11(21)10-23-16(22)9-12-5-2-3-8-15(12)20-17-13(18)6-4-7-14(17)19;1-11(10-21)23-16(22)9-12-5-2-3-8-15(12)20-17-13(18)6-4-7-14(17)19/h2*2-8,11,20-21H,9-10H2,1H3 |
| SMILES | CC(O)COC(=O)Cc1ccccc1Nc2c(Cl)cccc2Cl.CC(CO)OC(=O)Cc3ccccc3Nc4c(Cl)cccc4Cl |
| IUPAC Name | 1-hydroxypropan-2-yl 2-[2-(2,6-dichloroanilino)phenyl]acetate;2-hydroxypropyl 2-[2-(2,6-dichloroanilino)phenyl]acetate |
| Molecular Weight (g/mol) | 708.46 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction