missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Canadine, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Canadine
Specifications
Specifications
| Chemical Name or Material | Tetrahydroberberine (Canadine) |
| CAS | 522-97-4 |
| Analyte or Component Names | Tetrahydroberberine (Canadine) |
| Shipping Condition | Room Temperature |
| Molecular Formula | C20 H21 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 6H-Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizine, 5,8,13,13a-tetrahydro-9,10-dimethoxy-, Berberine, 9-deoxy-16,17-dihydro- (6CI), Berbine, 9,10-dimethoxy-2,3-(methylenedioxy)- (7CI,8CI), Berberine, tetrahydro-, Canadine, NSC 36351, NSC 94918, Tetrahydroberberine, Tetrahydroumbellatine, Xanthopuccine, dl-Canadine, dl-Tetrahydroberberine, (+/-)-Canadine, (+/-)-Tetrahydroberberine |
| InChI Formula | InChI=1S/C20H21NO4/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2/h3-4,8-9,16H,5-7,10-11H2,1-2H3 |
| SMILES | COc1ccc2CC3N(CCc4cc5OCOc5cc34)Cc2c1OC |
| Molecular Weight (g/mol) | 339.38 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction