missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Boldenone Undecylenate, Mikromol™
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
Description
Boldenone Undecylenate
Specifications
Specifications
| Chemical Name or Material | Boldenone Undecylenate |
| CAS | 13103-34-9 |
| Analyte or Component Names | Boldenone Undecylenate |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C30 H44 O3 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Ba 29038, Ba 9038, Boldefarm, Boldenone 10-undecenoate, Boldenone undecylenate, Equipoise, Parenabol, Vebonol, (17beta)-17-[(1-Oxo-10-undecenyl)oxy]-androsta-1,4-dien-3-one, Androsta-1,4-dien-3-one, 17beta-hydroxy-, 10-undecenoate (7CI,8CI), 10-Undecenoic acid, ester with 17beta-hydroxyandrosta-1,4-dien-3-one (8CI) |
| InChI Formula | InChI=1S/C30H44O3/c1-4-5-6-7-8-9-10-11-12-28(32)33-27-16-15-25-24-14-13-22-21-23(31)17-19-29(22,2)26(24)18-20-30(25,27)3/h4,17,19,21,24-27H,1,5-16,18,20H2,2-3H3/t24-,25-,26-,27-,29-,30-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](CCC4=CC(=O)C=C[C@]34C)[C@@H]1CC[C@@H]2OC(=O)CCCCCCCCC=C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17S)-10,13-dimethyl-3-oxo-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] undec-10-enoate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction