missing translation for 'onlineSavingsMsg'
Learn More
Learn More
10-[3-(Dimethyl-amino)propyl]acridin-9(10H)-one Hydrochloride, Mikromol™
Description
10-[3-(Dimethyl-amino)propyl]acridin-9(10H)-one Hydrochloride
Specifications
Specifications
| Chemical Name or Material | 10-[3-(Dimethyl-amino)propyl]acridin-9(10H)-one Hydrochloride |
| CAS | 2307-88-2 |
| Analyte or Component Names | 10-[3-(Dimethyl-amino)propyl]acridin-9(10H)-one Hydrochloride |
| Shipping Condition | Room Temperature |
| Molecular Formula | C18 H20 N2 O . Cl H |
| Certifications/Compliance | ISO 17025 |
| Synonym | Imipramine Hydrochloride Imp. C (EP), Imipramine Imp. C (EP), 10-[3-(Dimethyl-amino)propyl]acridin-9(10H)-one Hydrochloride, Imipramine Hydrochloride Imp. C (EP) as Hydrochloride, Imipramine Hydrochloride Impurity C as Hydrochloride, Imipramine Impurity C as Hydrochloride |
| InChI Formula | InChI=1S/C18H20N2O.ClH/c1-19(2)12-7-13-20-16-10-5-3-8-14(16)18(21)15-9-4-6-11-17(15)20;/h3-6,8-11H,7,12-13H2,1-2H3;1H |
| SMILES | Cl.CN(C)CCCN1c2ccccc2C(=O)c3ccccc13 |
| IUPAC Name | 10-[3-(dimethylamino)propyl]acridin-9-one;hydrochloride |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?