missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Neuron-specific beta-III Tubulin Alexa Fluor™ 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC1195V-100UG
This item is not returnable.
View return policy
Description
beta-III Tubulin Monoclonal antibody specifically detects beta-III Tubulin in Broad Species Reactivity samples. It is validated for Flow Cytometry
Specifications
| beta-III Tubulin | |
| Monoclonal | |
| Alexa Fluor 405 | |
| beta-4, TUBB4 CFEOM3A, tubulin beta-3 chain, Tubulin beta-4 chain, Tubulin beta-III, tubulin, beta 3, TUJ1 antigen, TUJ-1 antigen | |
| Rat brain-derived microtubules | |
| 100 μg | |
| Primary | |
| Broad Species Reactivity | |
| Purified |
| Flow Cytometry | |
| TuJ-1 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10381 | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2°C to 8°C as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction