Learn More
10-Hydroxy-10,11-dihydrocarbamazepine, Mikromol™
Product Code. 30459062
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30459062 4 x 25 mg 100mg
30050743 25 mg 25mg
17960406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30459062 Supplier Mikromol Supplier No. MM0076.1000254AMP
 Restricted Item
Estimated Shipment: 02-11-2026
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
10-Hydroxy-10,11-dihydrocarbamazepine, Mikromol
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10,11-Dihydro-10-Hydroxycarbamazepine
CAS 29331-92-8
Analyte or Component Names 10,11-Dihydro-10-Hydroxycarbamazepine
Shipping Condition Room Temperature
Molecular Formula C15 H14 N2 O2
Certifications/Compliance ISO 17025
Synonym 10-Hydroxy-10,11-dihydrocarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-hydroxycarbamazepine, 10-Hydroxy-10,11-dihydrocarbamezepine, BIA 2-005, GP 47779, Licarbazepine
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)
Solution Type Neat
SMILES NC(=O)N1c2ccccc2CC(O)c3ccccc13
IUPAC Name 5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Molecular Weight (g/mol) 254.28
Quantity 4 x 25 mg
Formula Weight 254.1055 g/mol