Learn More
10-Hydroxy-10,11-dihydrocarbamazepine, LoGiCal
Product Code. 30461298
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30461298 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30461298 Supplier LoGiCal Supplier No. LGCFOR0076.10
 Restricted Item
Estimated Shipment: 02-11-2026
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10,11-Dihydro-10-Hydroxycarbamazepine
CAS 29331-92-8
Analyte or Component Names 10,11-Dihydro-10-Hydroxycarbamazepine
Shipping Condition Room Temperature
Molecular Formula C15 H14 N2 O2
Certifications/Compliance ISO 9001
Synonym 10-Hydroxy-10,11-dihydrocarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-hydroxycarbamazepine, 10-Hydroxy-10,11-dihydrocarbamezepine, BIA 2-005, GP 47779, Licarbazepine
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)
Solution Type Neat
SMILES NC(=O)N1c2ccccc2CC(O)c3ccccc13
IUPAC Name 5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Industry Type Clinical Testing
Molecular Weight (g/mol) 254.28
Quantity 10 mg
Formula Weight 254.1055 g/mol