Learn More
10-Hydroxy-10,11-dihydrocarbamazepine 1.0 mg/ml in Acetonitrile, LoGiCal
Product Code. 30462737
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30462737 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30462737 Supplier LoGiCal Supplier No. LGCAMP0076.1011
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
LoGiCal's certified reference materials as neats, native solutions and stable isotope labelled solutions, with expert documentation and support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 10,11-Dihydro-10-Hydroxycarbamazepine
CAS 29331-92-8
Analyte or Component Names 10,11-Dihydro-10-Hydroxycarbamazepine
Shipping Condition Ice pack (-20°C)
Molecular Formula C15 H14 N2 O2
Certifications/Compliance ISO 17034
Synonym 10-Hydroxy-10,11-dihydrocarbamazepine, 10,11-Dihydro-10-hydroxy-5H-dibenz[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-hydroxycarbamazepine, 10-Hydroxy-10,11-dihydrocarbamezepine, BIA 2-005, GP 47779, Licarbazepine
InChI Formula InChI=1S/C15H14N2O2/c16-15(19)17-12-7-3-1-5-10(12)9-14(18)11-6-2-4-8-13(11)17/h1-8,14,18H,9H2,(H2,16,19)
Solution Type Single Solution
SMILES NC(=O)N1c2ccccc2CC(O)c3ccccc13
IUPAC Name 5-hydroxy-5,6-dihydrobenzo[b][1]benzazepine-11-carboxamide
Solvent or Matrix Acetonitrile
Industry Type Clinical Testing
Molecular Weight (g/mol) 254.28
Quantity 1 mL
Formula Weight 254.1055 g/mol