missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Piperonylpiperazine Dihydrochloride 1.0 mg/ml in Methanol (as free base), LoGiCal
Specifications
Specifications
| Chemical Name or Material | 1-Piperonylpiperazine Dihydrochloride |
| CAS | 38063-96-6 |
| Analyte or Component Names | 1-Piperonylpiperazine Dihydrochloride |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C12 H16 N2 O2 . 2 Cl H |
| Certifications/Compliance | ISO 17034 |
| Synonym | 1-(1,3-Benzodioxol-5-ylmethyl)piperazine Hydrochloride (1:2), Piperazine, 1-(1,3-benzodioxol-5-ylmethyl)-, dihydrochloride (9CI), Piperazine, 1-piperonyl-, dihydrochloride (7CI), 1-Piperonylpiperazine dihydrochloride |
| InChI Formula | InChI=1S/C12H16N2O2.2ClH/c1-2-11-12(16-9-15-11)7-10(1)8-14-5-3-13-4-6-14;;/h1-2,7,13H,3-6,8-9H2;2*1H |
| Solution Type | Single Solution |
| SMILES | Cl.Cl.C(N1CCNCC1)c2ccc3OCOc3c2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?