Learn More
1-Pentanesulfonic acid sodium salt, monohydrate; 99.9%, For HPLC analysis, MP Biomedicals™
Shop All MP Biomedicals ProductsSpecifications
Specifications
| Item Name | 1-Pentanesulfonic Acid, Sodium Salt, Monohydrate |
| Content And Storage | Store at Room Temperature. |
| Form | Crystalline Powder |
| Quantity | 5 g |
| Appearance | White |