missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Molecular weight: 398.57 g/mol
- Biological activity: Cell-permeable analog of the PKC-activating second messenger diacylglycerol (DAG); activates PKC in platelets, resulting in the phosphorylation of a 40 kDa protein
- Metabolism: Metabolized to its corresponding phosphatidic acid (1-oleoyl-2-acetyl-3-phosphoglycerol)
- Constituents: 100% 1-Oleoyl-2-acetyl-sn-glycerol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Buffer | 100% 1-Oleoyl-2-acetyl-sn-glycerol |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Lipid-based Substrate |
| Target Molecule | Protein Kinase C |
| Formulation | 100% 1-Oleoyl-2-acetyl-sn-glycerol |
| Quantity | 25 mg |
| For Use With (Application) | PKC activation |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?