Learn More
1-Octanesulfonic acid sodium salt, monohydrate, 99.7%, For HPLC analysis, MP Biomedicals™
Product Code. p-4404842 Shop All MP Biomedical Products
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. 17881371
Brand: MP Biomedicals 0215177805
Specifications
Item Name 1-Octanesulfonic Acid, Sodium Salt, Monohydrate
Content And Storage Store at room temperature (15°C to 30°C)
Form Crystalline Powder
Quantity 5 g
Appearance White