missing translation for 'onlineSavingsMsg'
Learn More

1-O-Benzyl 6-O-Tosyl-D-mannose, TRC

Product Code. 30336140
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30336140 500 mg 500mg
30345875 1 g 1g
30345793 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30336140 Supplier Toronto Research Chemicals Supplier No. TRCB170685500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1-O-Benzyl 6-O-Tosyl-D-mannose
Analyte or Component Names 1-O-Benzyl 6-O-Tosyl-D-mannose
Shipping Condition Room Temperature
Molecular Formula C20 H24 O8 S
Synonym ((2R,3S,4S,5S)-3,4,5-Trihydroxy-6-phenoxytetrahydro-2H-pyran-2-yl)methyl 4-Methylbenzenesulfonate
InChI Formula InChI=1S/C20H24O8S/c1-13-7-9-15(10-8-13)29(24,25)27-12-16-17(21)18(22)19(23)20(28-16)26-11-14-5-3-2-4-6-14/h2-10,16-23H,11-12H2,1H3/t16-,17-,18+,19+,20?/m1/s1
Solution Type Neat
SMILES Cc1ccc(cc1)S(=O)(=O)OC[C@H]2OC(OCc3ccccc3)[C@@H](O)[C@@H](O)[C@@H]2O
IUPAC Name [(2R,3S,4S,5S)-3,4,5-trihydroxy-6-phenylmethoxyoxan-2-yl]methyl 4-methylbenzenesulfonate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 424.465
Quantity 500 mg
Formula Weight 424.119
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.