Learn More
1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene, 99+%, Thermo Scientific™
Product Code. 11496180
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
Product Code. 11496180
Brand: Thermo Scientific Alfa Aesar H27102.09
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 85583-83-1
Molecular Formula C20H22O
Molecular Weight (g/mol) 278.395
MDL Number MFCD01218016
InChI Key ULERJJLPGXLNFM-UHFFFAOYSA-N
Synonym 1-n-butyl-4-4-ethoxyphenyl ethynyl benzene, 1-butyl-4-2-4-ethoxyphenyl ethynyl benzene, 1-2-4-butylphenyl ethynyl-4-ethoxybenzene, 1-butyl-4-4-ethoxyphenyl ethynyl benzene, 1-4-n-butylphenyl-2-4-ethoxyphenyl acetylene, 1-4-butylphenyl ethynyl-4-ethoxybenzene, 1-butyl-4-4-ethoxyphenyl ethynyl benzene #, benzene, 1-butyl-4-4-ethoxyphenyl ethynyl
PubChem CID 610253
IUPAC Name 1-butyl-4-[2-(4-ethoxyphenyl)ethynyl]benzene
SMILES CCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC
Specifications
Melting Point 54°C (N)
Boiling Point 168°C to 170°C (0.5 mmHg)
Quantity 10 g
Formula Weight 278.39
Percent Purity ≥99%
Chemical Name or Material 1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene