CAS RN 85583-83-1
CAS RN 85583-83-1
IUPAC Name:
1-butyl-4-[2-(4-ethoxyphenyl)ethynyl]benzene
Synonyms:
1-butyl-4-[2-(4-ethoxyphenyl)ethynyl]benzene
Molecular Weight (g/mol):
278.4
Molecular Formula:
C20H22O
InChi Key:
ULERJJLPGXLNFM-UHFFFAOYSA-N
SMILES:
CCCCC1=CC=C(C=C1)C#CC1=CC=C(OCC)C=C1
1
–
1
of
1
results
1
1-n-Butyl-4-[(4-ethoxyphenyl)ethynyl]benzene, 99+%, Thermo Scientific™
CAS: 85583-83-1 Molecular Formula: C20H22O Molecular Weight (g/mol): 278.395 MDL Number: MFCD01218016 InChI Key: ULERJJLPGXLNFM-UHFFFAOYSA-N Synonym: 1-n-butyl-4-4-ethoxyphenyl ethynyl benzene,1-butyl-4-2-4-ethoxyphenyl ethynyl benzene,1-2-4-butylphenyl ethynyl-4-ethoxybenzene,1-butyl-4-4-ethoxyphenyl ethynyl benzene,1-4-n-butylphenyl-2-4-ethoxyphenyl acetylene,1-4-butylphenyl ethynyl-4-ethoxybenzene,1-butyl-4-4-ethoxyphenyl ethynyl benzene #,benzene, 1-butyl-4-4-ethoxyphenyl ethynyl PubChem CID: 610253 IUPAC Name: 1-butyl-4-[2-(4-ethoxyphenyl)ethynyl]benzene SMILES: CCCCC1=CC=C(C=C1)C#CC2=CC=C(C=C2)OCC