missing translation for 'onlineSavingsMsg'
Learn More

1-Methyl-4-nitro-5-thioimidazole, TRC

Product Code. 30134645
Change view
Click to view available options
Quantity:
1 g
2.5 g
25 g
Unit Size:
1g
2.5g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134645 25 g 25g
30138523 2.5 g 2.5g
17914065 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30134645 Supplier Toronto Research Chemicals Supplier No. TRCM32468525G
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6339-54-4
Molecular Formula C4 H5 N3 O2 S
Molecular Weight (g/mol) 159.17
Synonym 1H-Imidazole-5-thiol, 1-methyl-4-nitro-, Imidazole-5-thiol, 1-methyl-4-nitro- (6CI,7CI,8CI), 1-Methyl-4-nitro-1H-imidazole-5-thiol, 1-Methyl-4-nitro-5-thioimidazole, 5-Mercapto-1-methyl-4-nitroimidazole
IUPAC Name 3-methyl-5-nitroimidazole-4-thiol
SMILES Cn1cnc(c1S)[N+](=O)[O-]

Specifications

Quantity 25 g
InChI Formula InChI=1S/C4H5N3O2S/c1-6-2-5-3(4(6)10)7(8)9/h2,10H,1H3
Formula Weight 159.0102
Chemical Name or Material 1-Methyl-4-nitro-5-thioimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.