CAS RN 6339-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-Methyl-4-nitro-5-thioimidazole, TRC
CAS: 6339-54-4 Molecular Formula: C4 H5 N3 O2 S Molecular Weight (g/mol): 159.17 Synonym: 1H-Imidazole-5-thiol, 1-methyl-4-nitro-,Imidazole-5-thiol, 1-methyl-4-nitro- (6CI,7CI,8CI),1-Methyl-4-nitro-1H-imidazole-5-thiol,1-Methyl-4-nitro-5-thioimidazole,5-Mercapto-1-methyl-4-nitroimidazole IUPAC Name: 3-methyl-5-nitroimidazole-4-thiol SMILES: Cn1cnc(c1S)[N+](=O)[O-]
2
1-Methyl-4-nitro-1H-imidazole-5-thiol, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.