Learn More
1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride, 97+%, Thermo Scientific™
Product Code. p-4509675
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10246532 250 mg Amber Glass Bottle 250mg
10397202 1 g Amber Glass Bottle 1g
11366815 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10246532 Supplier Thermo Scientific Maybridge Supplier No. AW00087CB
Add to basket
Chemical Identifiers
CAS 519056-67-8
Molecular Formula C5H4ClF3N2O2S
Molecular Weight (g/mol) 248.604
InChI Key HJFWSNIHZZKYIK-UHFFFAOYSA-N
Synonym 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl pyrazole-4-sulfonyl chloride, 1h-pyrazole-4-sulfonylchloride,1-methyl-3-trifluoromethyl, chloro 1-methyl-3-trifluoromethyl pyrazol-4-yl sulfone, 1-methyl-3-trifluoromethyl-1h-pyrazole-4-sulfonylchloride, 1h-pyrazole-4-sulfonyl chloride, 1-methyl-3-trifluoromethyl
PubChem CID 2794561
IUPAC Name 1-methyl-3-(trifluoromethyl)pyrazole-4-sulfonyl chloride
SMILES CN1C=C(C(=N1)C(F)(F)F)S(=O)(=O)Cl
Specifications
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 248.61
Percent Purity 97%
Chemical Name or Material 1-Methyl-3-(trifluoromethyl)-1H-pyrazole-4-sulfonyl chloride