missing translation for 'onlineSavingsMsg'
Learn More

1-Hydroxy Ibuprofen (Ibuprofen Impurity L) (Mixture of Diastereomers), TRC

Product Code. 30004793
Change view
Click to view available options
Quantity:
5 mg
25 mg
50 mg
Unit Size:
25mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004793 50 mg 50mg
17981725 25 mg 25mg
17910695 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30004793 Supplier Toronto Research Chemicals Supplier No. TRCH94285850MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53949-53-4
Molecular Formula C13 H18 O3
Molecular Weight (g/mol) 222.28
Synonym Benzeneacetic acid, 4-(1-hydroxy-2-methylpropyl)-α-methyl-, 1-Hydroxyibuprofen, 4-(1-Hydroxy-2-methylpropyl)-α-methylbenzeneacetic acid, 2-[4'-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid
IUPAC Name 2-[4-(1-hydroxy-2-methylpropyl)phenyl]propanoic acid
SMILES CC(C)C(O)c1ccc(cc1)C(C)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H18O3/c1-8(2)12(14)11-6-4-10(5-7-11)9(3)13(15)16/h4-9,12,14H,1-3H3,(H,15,16)
Formula Weight 222.1256
Percent Purity >95%
Chemical Name or Material 2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (1-Hydroxyibuprofen)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.