CAS RN 53949-53-4
CAS RN 53949-53-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
2-[4-(1-Hydroxy-2-methylpropyl)phenyl]propanoic Acid (1-Hydroxyibuprofen), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
1-Hydroxy Ibuprofen (Ibuprofen Impurity L) (Mixture of Diastereomers), TRC
CAS: 53949-53-4 Molecular Formula: C13 H18 O3 Molecular Weight (g/mol): 222.28 Synonym: Benzeneacetic acid, 4-(1-hydroxy-2-methylpropyl)-α-methyl-,1-Hydroxyibuprofen,4-(1-Hydroxy-2-methylpropyl)-α-methylbenzeneacetic acid,2-[4'-(1-Hydroxy-2-methylpropyl)phenyl]propionic acid IUPAC Name: 2-[4-(1-hydroxy-2-methylpropyl)phenyl]propanoic acid SMILES: CC(C)C(O)c1ccc(cc1)C(C)C(=O)O