Learn More
(1-Butyl)triphenylphosphonium chloride
Product Code. 11475500
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. 11475500
Brand: Thermo Scientific Alfa Aesar H25796.22
Request Bulk or Custom Format
CAS: 13371-17-0 | C22H24ClP | 354.86 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13371-17-0
Molecular Formula C22H24ClP
Molecular Weight (g/mol) 354.86
MDL Number MFCD00040546
InChI Key MFIUDWFSVDFDDY-UHFFFAOYSA-M
Synonym butyltriphenylphosphonium chloride, butyl triphenyl phosphonium chloride, phosphonium, butyltriphenyl-, chloride, 1-butyl triphenylphosphonium chloride, phosphonium, butyltriphenyl-, chloride 1:1, butyltriphenyl phosphonium chloride, butyl triphenyl phosphanium chloride, butyltriphenylphosphanium chloride, acmc-1bzbo, butyltriphenyl-phosphoniuchloride
PubChem CID 166797
IUPAC Name butyl(triphenyl)phosphanium;chloride
SMILES [Cl-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Specifications
Melting Point 225°C to 228°C
Linear Formula CH3(CH2)3P(C6H5)3Cl
Quantity 100 g
UN Number UN3464
Sensitivity Hygroscopic
Formula Weight 354.86
Chemical Name or Material (1-Butyl)triphenylphosphonium chloride