accessibility menu, dialog, popup

UserName

CAS RN 13371-17-0

ClH3CP

CAS RN 13371-17-0

IUPAC Name: butyltriphenylphosphanium chloride
Synonyms: butyltriphenylphosphanium chloride
Molecular Weight (g/mol): 354.86
Molecular Formula: C22H24ClP
InChi Key: MFIUDWFSVDFDDY-UHFFFAOYSA-M
SMILES: [Cl-].CCCC[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results