Learn More
1-Acetyl-5-bromo-7-nitroindoline, 98%, Thermo Scientific™
Product Code. 11334955
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334955 5 g 5g
11324955 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11334955 Supplier Thermo Scientific Alfa Aesar Supplier No. L18271.06
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 62368-07-4
Molecular Formula C10H9BrN2O3
Molecular Weight (g/mol) 285.10
MDL Number MFCD00056018
InChI Key RCELVCGNAKOBPO-UHFFFAOYSA-N
Synonym 1-acetyl-5-bromo-7-nitroindoline, 1-5-bromo-7-nitroindolin-1-yl ethanone, n-acetyl-5-bromo-7-nitro-indoline, 1-acetyl-5-bromo-7-nitroindole, 1-5-bromo-7-nitro-2,3-dihydroindol-1-yl ethanone, 1-acetyl-5-bromo-7-nitro-2,3-dihydro-1h-indole, pubchem7303, acmc-1bfxp, n-acetyl-5-bromo-7-nitroindoline
PubChem CID 2729243
IUPAC Name 1-(5-bromo-7-nitro-2,3-dihydroindol-1-yl)ethanone
SMILES CC(=O)N1CCC2=CC(Br)=CC(=C12)[N+]([O-])=O
Specifications
Melting Point 196°C to 198°C
Quantity 5 g
Beilstein 237670
Formula Weight 285.1
Percent Purity 98%
Chemical Name or Material 1-Acetyl-5-bromo-7-nitroindoline