missing translation for 'onlineSavingsMsg'
Learn More

1-Acetoxy-4-diethylamino-2-butyne, 98%, Thermo Scientific™

Product Code. 15420418
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15420418 25 g 25g
15410418 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15420418 Supplier Thermo Scientific Alfa Aesar Supplier No. H53511.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22396-77-6
Molecular Formula C10H17NO2
Molecular Weight (g/mol) 183.25
MDL Number MFCD00190224
InChI Key AQEDWWHCJSXKHP-UHFFFAOYSA-N
Synonym 1-acetoxy-4-diethylamino-2-butyne, 4-diethylamino but-2-yn-1-yl acetate, 4-diethylamino-but-2-yn-1-yl acetate, 4-diethylamino but-2-ynyl acetate, acmc-209up2, acetic acid 4-diethylamino-2-butynyl ester, 2-butyn-1-ol,4-diethylamino-acetate ester
PubChem CID 2756256
IUPAC Name 4-(diethylamino)but-2-ynyl acetate
SMILES CCN(CC)CC#CCOC(C)=O

Specifications

Boiling Point 217°C to 218°C
Flash Point 103°C (217°F)
Linear Formula CH3CO2CH2C≡CCH2N(CH2CH3)2
Refractive Index 1.456
Quantity 25 g
Formula Weight 183.25
Percent Purity 98%
Chemical Name or Material 1-Acetoxy-4-diethylamino-2-butyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.