accessibility menu, dialog, popup

UserName

CH3NH3COH3CO

CAS RN 22396-77-6

IUPAC Name: 4-(diethylamino)but-2-yn-1-yl acetate
Synonyms: 4-(diethylamino)but-2-yn-1-yl acetate
Molecular Weight (g/mol): 183.25
Molecular Formula: C10H17NO2
InChi Key: AQEDWWHCJSXKHP-UHFFFAOYSA-N
SMILES: CCN(CC)CC#CCOC(C)=O
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results