missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromoperfluorohexane, 96%, Thermo Scientific™

Product Code. 11469737
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469737 1 g 1g
11479737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469737 Supplier Thermo Scientific Alfa Aesar Supplier No. B22729.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Solubility
Soluble in water.

Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 918-22-9
Molecular Formula C6Br2F12
Molecular Weight (g/mol) 459.86
MDL Number MFCD00153110
InChI Key SWAPKQWKFYIOJS-UHFFFAOYSA-N
Synonym 1,6-dibromododecafluorohexane, 1,6-dibromoperfluorohexane, hexane, 1,6-dibromo-dodecafluoro, dibromoperfluorohexane, acmc-209rdr, perfluoro 1,6-dibromohexane, hexane,1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluoro, 1,6-bis bromanyl-1,1,2,2,3,3,4,4,5,5,6,6-dodecakis fluoranyl hexane
PubChem CID 522992
IUPAC Name 1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexane
SMILES FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

Specifications

Boiling Point 141°C to 142°C
Refractive Index 1.345
Quantity 1 g
Beilstein 1822184
Solubility Information Soluble in water.
Formula Weight 459.87
Percent Purity 96%
Chemical Name or Material 1,6-Dibromoperfluorohexane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.