accessibility menu, dialog, popup

UserName

FFBrFFFFFFBrFFFF

CAS RN 918-22-9

IUPAC Name: 1,6-dibromo-1,1,2,2,3,3,4,4,5,5,6,6-dodecafluorohexane
Synonyms: 1,6-dibromododecafluorohexane
Molecular Weight (g/mol): 459.86
Molecular Formula: C6Br2F12
InChi Key: SWAPKQWKFYIOJS-UHFFFAOYSA-N
SMILES: FC(F)(Br)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)