Learn More
1,4-Diethoxybenzene, 98%
Product Code. 11438336
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11438336 25 g 25g
11448336 100 g 100g
11458336 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11438336 Supplier Thermo Scientific Alfa Aesar Supplier No. A15385.14
Request Bulk or Custom Format
CAS: 122-95-2 | C10H14O2 | 166.22 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 122-95-2
Molecular Formula C10H14O2
Molecular Weight (g/mol) 166.22
MDL Number MFCD00015146
InChI Key VWGNFIQXBYRDCH-UHFFFAOYSA-N
Synonym benzene, 1,4-diethoxy, p-diethoxybenzene, hydroquinone diethyl ether, benzene, p-diethoxy, diethoxybenzene, 1,4-diethoxy-benzen, benzene,4-diethoxy, 1,4-diethoxybenzol, hydroquinonedlethylether, acmc-209ans
PubChem CID 67150
IUPAC Name 1,4-diethoxybenzene
SMILES CCOC1=CC=C(OCC)C=C1
Specifications
Melting Point 69°C to 71°C
Boiling Point 245°C to 248°C
Quantity 25 g
Beilstein 2044373
Formula Weight 166.22
Percent Purity 98%
Chemical Name or Material 1,4-Diethoxybenzene