CAS RN 122-95-2
IUPAC Name:
1,4-diethoxybenzene
Synonyms:
diethoxybenzene
Molecular Weight (g/mol):
166.22
Molecular Formula:
C10H14O2
InChi Key:
VWGNFIQXBYRDCH-UHFFFAOYSA-N
SMILES:
CCOC1=CC=C(OCC)C=C1
1
–
1
of
1
results
1
1,4-Diethoxybenzene, 98%
CAS: 122-95-2 Molecular Formula: C10H14O2 Molecular Weight (g/mol): 166.22 MDL Number: MFCD00015146 InChI Key: VWGNFIQXBYRDCH-UHFFFAOYSA-N Synonym: benzene, 1,4-diethoxy,p-diethoxybenzene,hydroquinone diethyl ether,benzene, p-diethoxy,diethoxybenzene,1,4-diethoxy-benzen,benzene,4-diethoxy,1,4-diethoxybenzol,hydroquinonedlethylether,acmc-209ans PubChem CID: 67150 IUPAC Name: 1,4-diethoxybenzene SMILES: CCOC1=CC=C(OCC)C=C1