Learn More
Trajan Scientific and Medical™ 1/32 in. O.D. PEEKsil™ Tubing
Product Code. 15208763
Click to view available options
Overall Length (Metric):
50 mm
100 mm
150 mm
250 mm
500 mm
Diameter (Metric) Inner:
0.025 mm
0.05 mm
0.075 mm
0.1 mm
0.15 mm
Unit Size:
Each
Pack of 2
Product Code. 15208763
Brand: Trajan Scientific and Medical™ 0624265
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
A mechanically robust polymer-sheathed fused silica tubing for use in HPLC systems
  • Fused silica internal wall provides smooth surface, free of imperfections
  • PEEKsil compatible with most organic solvents and resistant to strong acids
  • Effective pH range of 0 to 10
  • Flexibility ensures easy connection between columns, detectors and injectors
    Specifications
    Type Tubing
    Includes 2/Pk.
    Diameter (Metric) Inner 0.05 mm
    Overall Length (Metric) 250 mm