missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,3-Di-n-butylxanthine, 98%

Product Code. 11464487
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464487 25 g 25g
11474487 100 g 100g
11484487 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464487 Supplier Alfa Aesar™ Supplier No. B20491.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 2850-36-4
Molecular Formula C13H20N4O2
Molecular Weight (g/mol) 264.329
MDL Number MFCD00798591
InChI Key HDAPVDGMACVEKL-UHFFFAOYSA-N
Synonym 1,3-dibutylxanthine, 1,3-di-n-butylxanthine, 1,3-dibutyl-1h-purine-2,6 3h,7h-dione, 1,3-dibutyl-3,7-dihydro-1h-purine-2,6-dione, unii-ir9d2ugy27, ir9d2ugy27, 1,3-dibutyl-3,7-dihydro-purine-2,6-dione, acmc-20a2rg, 1,3-di-n-butyl-xanthine, 1,3-di-n-butyl xanthine
PubChem CID 101421
IUPAC Name 1,3-dibutyl-7H-purine-2,6-dione
SMILES CCCCN1C2=C(C(=O)N(C1=O)CCCC)NC=N2

Specifications

Quantity 25 g
Formula Weight 264.33
Percent Purity 98%
Chemical Name or Material 1,3-Di-n-butylxanthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.