missing translation for 'onlineSavingsMsg'
Learn More

1-(3-Chlorophenyl)-1,2-propanedione (>85%), TRC

Product Code. 30010444
Change view
Click to view available options
Quantity:
25 mg
100 mg
1 g
Unit Size:
100mg
1g
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010444 1 g 1g
30009602 100 mg 100mg
30009664 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010444 Supplier Toronto Research Chemicals Supplier No. TRCC3796451G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10557-17-2
Molecular Formula C9H7ClO2
Molecular Weight (g/mol) 182.6
Synonym 1-(3-Chlorophenyl)-1,2-propanedione, 1,2-Propanedione, 1-(m-chlorophenyl)- (7CI,8CI)
IUPAC Name 1-(3-chlorophenyl)propane-1,2-dione
SMILES CC(=O)C(=O)c1cccc(Cl)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H7ClO2/c1-6(11)9(12)7-3-2-4-8(10)5-7/h2-5H,1H3
Formula Weight 182.0135
Chemical Name or Material 1-(3-Chlorophenyl)-1,2-propanedione
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.