CAS RN 10557-17-2
CAS RN 10557-17-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
1-(3-Chlorophenyl)-1,2-propandione, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
1-(3-Chlorophenyl)-1,2-propanedione (>85%), TRC
CAS: 10557-17-2 Molecular Formula: C9H7ClO2 Molecular Weight (g/mol): 182.6 Synonym: 1-(3-Chlorophenyl)-1,2-propanedione,1,2-Propanedione, 1-(m-chlorophenyl)- (7CI,8CI) IUPAC Name: 1-(3-chlorophenyl)propane-1,2-dione SMILES: CC(=O)C(=O)c1cccc(Cl)c1