Learn More
1,3,5-Tris(4-aminophenoxy)benzene, TRC
Product Code. 30451731
Change view
Click to view available options
Quantity:
500 mg
5 g
Unit Size:
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30451731 5 g 5g
30455880 500 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30451731 Supplier Toronto Research Chemicals Supplier No. TRCT8750055G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 102852-92-6
Molecular Formula C24H21N3O3
Molecular Weight (g/mol) 399.44
Synonym 4,4',4' '-[1,3,5-Benzenetriyltris(oxy)]trisbenzenamine, 1,3,5-Tris(p-aminophenoxy)benzene, 135TAPOB
IUPAC Name 4-[3,5-bis(4-aminophenoxy)phenoxy]aniline
SMILES Nc1ccc(Oc2cc(Oc3ccc(N)cc3)cc(Oc4ccc(N)cc4)c2)cc1
Specifications
Quantity 5 g
InChI Formula InChI=1S/C24H21N3O3/c25-16-1-7-19(8-2-16)28-22-13-23(29-20-9-3-17(26)4-10-20)15-24(14-22)30-21-11-5-18(27)6-12-21/h1-15H,25-27H2
Formula Weight 399.1583
Chemical Name or Material 1,3,5-Tris(4-aminophenoxy)benzene