Learn More
1,1-Dimethylethyl 2-[(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate, Mikromol™
Description
1,1-Dimethylethyl 2-[(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate
Specifications
Specifications
| Chemical Name or Material | 1,1-Dimethylethyl 2-[(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate |
| CAS | 125971-94-0 |
| Analyte or Component Names | 1,1-Dimethylethyl 2-[(4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate |
| Shipping Condition | Room Temperature |
| Molecular Formula | C14 H23 N O4 |
| Certifications/Compliance | ISO 17025 |
| Synonym | 1,3-Dioxane-4-acetic acid, 6-(cyanomethyl)-2,2-dimethyl-, 1,1-dimethylethyl ester, (4R,6R)-, (4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-1,3-dioxane-4-acetic Acid 1,1-Dimethylethyl Ester, : 1,3-Dioxane-4-acetic acid, 6-(cyanomethyl)-2,2-dimethyl-, 1,1-dimethylethyl ester, (4R-cis)-, ((4R,6R)-6-(Cyanomethyl)-2,2-dimethyl-[1,3]dioxan-4-yl)acetic acid tert-butyl ester, (6R-Cyanomethyl-2,2-dimethyl-[1,3]dioxan-4R-yl)acetic acid tert-butyl ester |
| InChI Formula | InChI=1S/C14H23NO4/c1-13(2,3)19-12(16)9-11-8-10(6-7-15)17-14(4,5)18-11/h10-11H,6,8-9H2,1-5H3/t10-,11-/m1/s1 |
| SMILES | CC(C)(C)OC(=O)C[C@H]1C[C@@H](CC#N)OC(C)(C)O1 |
| IUPAC Name | tert-butyl 2-[(4R,6R)-6-(cyanomethyl)-2,2-dimethyl-1,3-dioxan-4-yl]acetate |
| Show More |